MilliporeSigma
Protein separations using enhanced-fluidity liquid chromatography.
Journal of chromatography. A (2017-08-02)
Raffeal Bennett, Susan V Olesik
ABSTRACT
Enhanced-fluidity liquid chromatography (EFLC) methods using methanol/H
Sigma-Aldrich
SigmaProt Intact Protein LC-MS Standard, Proteomics Retention Time Standard