MilliporeSigma
  • Mechanism of Phospholipid Hydrolysis for Oyster Crassostrea plicatula Phospholipids During Storage Using Shotgun Lipidomics.
Mechanism of Phospholipid Hydrolysis for Oyster Crassostrea plicatula Phospholipids During Storage Using Shotgun Lipidomics.
Lipids (2017-10-05)
Qinsheng Chen, Xincen Wang, Peixu Cong, Yanjun Liu, Yuming Wang, Jie Xu, Changhu Xue
ABSTRACT
A fast and efficient shotgun lipidomics strategy was applied to analyze phospholipids (PL) in the oyster Crassostrea plicatula, including 29 species of phosphatidylcholine (PtdCho), 23 species of phosphatidylethanolamine (PtdEtn), 11 species of phosphatidylserine (PtdSer), 6 species of phosphatidylinositol (PtdIns), and 17 species of lysophospholipids (Lyso-PL). During storage at 4 °C for 7 days, the PL content decreased by 68.08%, but a significant increase in the FFA content was observed (from 63.11 to 318.72 μg/g). PtdCho and PtdIns decreased relatively by 64.97 and 67.49%, and PtdSer decreased most markedly by 74.15%. However, the PtdEtn content increased slightly during the early stages of storage but subsequently began to decrease. Moreover, PL with eicosapentaenoic acid (EPA-PL) and docosahexaenoic acid (DHA-PL) decreased by 51.77 and 50.61%, whereas plasmalogens were relatively stable showing only a 25.46% decrease. In particular, through enzyme activity analysis of lipase, phospholipase A
Sigma-Aldrich
4-Nitrophenyl palmitate, lipase substrate
Sigma-Aldrich
O-(4-Nitrophenylphosphoryl)choline