MilliporeSigma
  • Suppression of SRCAP chromatin remodelling complex and restriction of lymphoid lineage commitment by Pcid2.
Suppression of SRCAP chromatin remodelling complex and restriction of lymphoid lineage commitment by Pcid2.
Nature communications (2017-11-16)
Buqing Ye, Benyu Liu, Liuliu Yang, Guanling Huang, Lu Hao, Pengyan Xia, Shuo Wang, Ying Du, Xiwen Qin, Pingping Zhu, Jiayi Wu, Nobuo Sakaguchi, Junyan Zhang, Zusen Fan
ABSTRACT
Lymphoid lineage commitment is an important process in haematopoiesis, which forms the immune system to protect the host from pathogen invasion. However, how multipotent progenitors (MPP) switch into common lymphoid progenitors (CLP) or common myeloid progenitors (CMP) during this process remains elusive. Here we show that PCI domain-containing protein 2 (Pcid2) is highly expressed in MPPs. Pcid2 deletion in the haematopoietic system causes skewed lymphoid lineage specification. In MPPs, Pcid2 interacts with the Zinc finger HIT-type containing 1 (ZNHIT1) to block Snf2-related CREBBP activator protein (SRCAP) activity and prevents the deposition of histone variant H2A.Z and transcription factor PU.1 to key lymphoid fate regulator genes. Furthermore, Znhit1 deletion also abrogates H2A/H2A.Z exchange in MPPs. Thus Pcid2 controls lymphoid lineage commitment through the regulation of SRCAP remodelling activity.
Sigma-Aldrich
Nuclei Isolation Kit: Nuclei EZ Prep, sufficient for 25 nuclei preparations (~1-10×107 cells/preparation)
Pricing and availability is not currently available.
Sigma-Aldrich
ATPase/GTPase Activity Assay Kit, sufficient for 200 colorimetric tests
Pricing and availability is not currently available.