MilliporeSigma
  • Nose to Brain Delivery of Rivastigmine by In Situ Gelling Cationic Nanostructured Lipid Carriers: Enhanced Brain Distribution and Pharmacodynamics.
Nose to Brain Delivery of Rivastigmine by In Situ Gelling Cationic Nanostructured Lipid Carriers: Enhanced Brain Distribution and Pharmacodynamics.
Journal of pharmaceutical sciences (2017-09-20)
Preeti Wavikar, Rohan Pai, Pradeep Vavia
ABSTRACT
Present investigation explores the potential of nanostructured lipid carriers (NLCs) for nose to brain delivery of rivastigmine (RV), which is further enhanced by incorporating into an in situ gelling system, increasing retention in nasal cavity. NLCs having particle size of 123.2 ± 2.3 nm, entrapment efficiency of 68.3 ± 3.4%, and zeta potential of 32 ± 1.2 mV was fabricated by a scalable method. Pharmacokinetics showed sustained release of intranasal (IN) and intravenous (IV) NLCs compared with RV solution by same route, with significantly higher AUC and T
Sigma-Aldrich
1-Octanoyl-rac-glycerol, ≥99%