MilliporeSigma
  • Multifunctionalized mesoporous silica as an efficient reversed-phase/anion exchange mixed-mode sorbent for solid-phase extraction of four acidic nonsteroidal anti-inflammatory drugs in environmental water samples.
Multifunctionalized mesoporous silica as an efficient reversed-phase/anion exchange mixed-mode sorbent for solid-phase extraction of four acidic nonsteroidal anti-inflammatory drugs in environmental water samples.
Journal of chromatography. A (2017-11-06)
Yun Li, Chaonan Huang, Jiajia Yang, Junyu Peng, Jing Jin, Huilian Ma, Jiping Chen
ABSTRACT
A mesoporous silica Santa Barbara Amorphous-15 (SBA-15) has been first functionalized with 3-[2-(2-aminoethylamino)ethylamino]propyl-trimethoxysilane (a silane with three amines) and then reacted with an excess of phenyl glycidyl ether to generate a mixed-mode anion-exchanger containing both anion-exchange (three amines) and reversed-phase (multiple ether-linked phenyls) functionalities in a single branched ligand. The resulting material has been characterized by scanning electron microscopy, transmission electron microscopy, nitrogen adsorption-desorption measurements, Fourier-transform infrared spectroscopy, and elemental analysis. The results obtained indicated a BET specific surface area (S
Sigma-Aldrich
N1-(3-Trimethoxysilylpropyl)diethylenetriamine, technical grade
Sigma-Aldrich
DIC, purum, ≥98.0% (GC)