MilliporeSigma
  • DELLA1-Mediated Gibberellin Signaling Regulates Cytokinin-Dependent Symbiotic Nodulation.
DELLA1-Mediated Gibberellin Signaling Regulates Cytokinin-Dependent Symbiotic Nodulation.
Plant physiology (2017-10-20)
Camille Fonouni-Farde, Anna Kisiala, Mathias Brault, R J Neil Emery, Anouck Diet, Florian Frugier
ABSTRACT
In legume plants, low-nitrogen soils promote symbiotic interactions with rhizobial bacteria, leading to the formation of nitrogen-fixing root nodules. Among critical signals regulating this developmental process are bacterial Nod Factors (NFs) and several plant hormones, including cytokinins (CKs) and gibberellins (GAs). Here, we show in
Sigma-Aldrich
1-Formylpiperidine, 99%