MilliporeSigma
Histone propionylation is a mark of active chromatin.
Nature structural & molecular biology (2017-10-24)
Adam F Kebede, Anna Nieborak, Lara Zorro Shahidian, Stephanie Le Gras, Florian Richter, Diana Aguilar Gómez, Marijke P Baltissen, Gergo Meszaros, Helena de Fatima Magliarelli, Aaron Taudt, Raphael Margueron, Maria Colomé-Tatché, Romeo Ricci, Sylvain Daujat, Michiel Vermeulen, Gerhard Mittler, Robert Schneider
ABSTRACT
Histones are highly covalently modified, but the functions of many of these modifications remain unknown. In particular, it is unclear how histone marks are coupled to cellular metabolism and how this coupling affects chromatin architecture. We identified histone H3 Lys14 (H3K14) as a site of propionylation and butyrylation in vivo and carried out the first systematic characterization of histone propionylation. We found that H3K14pr and H3K14bu are deposited by histone acetyltransferases, are preferentially enriched at promoters of active genes and are recognized by acylation-state-specific reader proteins. In agreement with these findings, propionyl-CoA was able to stimulate transcription in an in vitro transcription system. Notably, genome-wide H3 acylation profiles were redefined following changes to the metabolic state, and deletion of the metabolic enzyme propionyl-CoA carboxylase altered global histone propionylation levels. We propose that histone propionylation, acetylation and butyrylation may act in combination to promote high transcriptional output and to couple cellular metabolism with chromatin structure and function.
Sigma-Aldrich
Histone from calf thymus, Type II-A, lyophilized powder