MilliporeSigma
  • Chronic irradiation with low-dose-rate 137Cs-γ rays inhibits NGF-induced neurite extension of PC12 cells via Ca2+/calmodulin-dependent kinase II activation.
Chronic irradiation with low-dose-rate 137Cs-γ rays inhibits NGF-induced neurite extension of PC12 cells via Ca2+/calmodulin-dependent kinase II activation.
Journal of radiation research (2017-11-07)
Katoh Shinsuke, Kobayashi Junya, Umeda Tomonobu, Kobayashi Yoshiko, Nobuo Izumo, Suzuki Takahiko
ABSTRACT
Chronic irradiation with low-dose-rate 137Cs-γ rays inhibits the differentiation of human neural progenitor cells and influences the expression of proteins associated with several cellular functions. We aimed to determine whether such chronic irradiation influences the expression of proteins associated with PC12 cells. Chronic irradiation at 0.027 mGy/min resulted in inhibition of NGF-induced neurite extension. Furthermore, irradiation enhanced the nerve growth factor (NGF)-induced increase in the phosphorylation of extracellular signal-regulated kinase (ERK), but did not affect the phosphorylation of NGF receptors, suggesting that irradiation influences pathways unassociated with the activation of ERK. We then examined whether irradiation influenced the Akt-Rac1 pathway, which is unaffected by ERK activation. Chronic irradiation also enhanced the NGF-induced increase in Akt phosphorylation, but markedly inhibited the NGF-induced increase in Rac1 activity that is associated with neurite extension. These results suggest that the inhibitory effect of irradiation on neurite extension influences pathways unassociated with Akt activation. As Ca2+/calmodulin-dependent kinase II (CaMKII) is known to inhibit the NGF-induced neurite extension in PC12 cells, independent of ERK and Akt activation, we next examined the effects of irradiation on CaMKII activation. Chronic irradiation induced CaMKII activation, while application of KN-62 (a specific inhibitor of CaMKII), attenuated increases in CaMKII activation and recovered neurite extension and NGF-induced increases in Rac1 activity that was inhibited by irradiation. Our results suggest that chronic irradiation with low-dose-rate γ-rays inhibits Rac1 activity via CaMKII activation, thereby inhibiting NGF-induced neurite extension.
Sigma-Aldrich
KN-62, ≥95%, powder