MilliporeSigma
Construction of a lateral flow strip for detection of soymilk in milk.
Journal of food science and technology (2017-12-01)
Priyae Brath Gautam, Rajan Sharma, Kiran Lata, Y S Rajput, Bimlesh Mann
ABSTRACT
A lateral flow based detection method for ascertaining the presence of soymilk in whole bovine milk has been described. The method uses commercially available rabbit anti-soy protein antibodies conjugated to gold nanoparticles (AuNPs) wherein soymilk protein in adulterated milk and soymilk protein at test line competes for limited antibodies. At control line, anti-rabbit immunoglobulin was immobilized for ensuring flow properties of antibody-conjugated AuNPs. Absence or diminished intensity of band at test line indicates presence of soymilk in milk. The soymilk detection limit was 1.75% (v/v) in whole bovine milk and results are available in 5 min. Constructed lateral flow device can be used for on-spot examination of soymilk in milk.
Sigma-Aldrich
Anti-Soy Protein antibody produced in rabbit, fractionated antiserum, buffered aqueous solution