MilliporeSigma
  • Ephemeral biogels to control anti-biofilm agent delivery: From conception to the construction of an active dressing.
Ephemeral biogels to control anti-biofilm agent delivery: From conception to the construction of an active dressing.
Materials science & engineering. C, Materials for biological applications (2017-10-14)
Elodie Lefebvre, Pierre Lembre, Julien Picard, Lamia El-Guermah, Damien Seyer, Véronique Larreta Garde
ABSTRACT
Chronic wound colonization by bacterial biofilms is common and can cause various complications. An anti-biofilm strategy was developed around the co-entrapment of a commercially available antiseptic, PHMB (polyhexamethylene biguanide 4mgmL
Sigma-Aldrich
Protease from Bacillus sp., liquid, ≥8 U/g