MilliporeSigma
  • Small peptide inhibitor of JNKs protects against MPTP-induced nigral dopaminergic injury via inhibiting the JNK-signaling pathway.
Small peptide inhibitor of JNKs protects against MPTP-induced nigral dopaminergic injury via inhibiting the JNK-signaling pathway.
Laboratory investigation; a journal of technical methods and pathology (2009-12-17)
Jing Pan, Jing Qian, Yu Zhang, Jianfang Ma, Gang Wang, Qin Xiao, Shengdi Chen, Jianqing Ding
ABSTRACT
Increasing evidence suggests that apoptosis may be the mechanism underlying cell death in selective loss of nigral dopaminergic neurons in Parkinson's disease (PD). Previous studies strongly suggested that c-Jun N-terminal kinase (JNK) signaling pathway has a critical role in the animal model with 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP)-induced PD. In this study, we report the inhibitory effect of a peptide designated as Tat-JBD on JNKs activation. The sequence of Tat is corresponding to the cell-membrane transduction domain of human immunodeficiency virus-type 1 (HIV-1) and the sequence of an 11-amino acid peptide is corresponding to the residues of JNK-binding domain (JBD) on JNK-interacting protein-1 (JIP-1). Tat-JBD is confirmed to perturb the assembly of JIP-1-JNKs complex, inhibit the activation of JNKs induced by MPTP and consequently diminish the phosphorylation of c-Jun. It also inhibits the phosphorylation of Bcl-2 and the releasing of Bax from Bcl-2/Bax dimmers, sequentially attenuates the translocation of Bax to mitochondria, the release of cytochrome c, the activation of caspase3 and the hydrolyzation of poly-ADP-ribose-polymerase. The death of dopaminergic neurons and the loss of dopaminergic axon in the striatum were significantly suppressed by infusion of the peptide Tat-JBD in MPTP-treated mice. Our findings imply that Tat-JBD offers neuroprotection against MPTP injury via inhibiting the JNK-signaling pathway, and may provide a promising therapeutic approach for PD.
Sigma-Aldrich
ApopTag Peroxidase In Situ Apoptosis Detection Kit, The ApopTag Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells in situ by labeling & detecting DNA strand breaks by the TUNEL method.