MilliporeSigma
Re-creation of a Key Step in the Evolutionary Switch from C
Current biology : CB (2017-10-24)
Peng Wang, Roxana Khoshravesh, Shanta Karki, Ronald Tapia, C Paolo Balahadia, Anindya Bandyopadhyay, W Paul Quick, Robert Furbank, Tammy L Sage, Jane A Langdale
ABSTRACT
The C
Product Description
Millipore
Universal Food pathogen ID Membrane, light pink colored, sterile, diameter 70 mm, suitable for microbiology