MilliporeSigma
  • Arctic ground squirrel resist peroxynitrite-mediated cell death in response to oxygen glucose deprivation.
Arctic ground squirrel resist peroxynitrite-mediated cell death in response to oxygen glucose deprivation.
Free radical biology & medicine (2017-10-01)
Saurav Bhowmick, Kelly L Drew
ABSTRACT
Cerebral ischemia-reperfusion (I/R) injury initiates a cascade of events, generating nitric oxide (NO) and superoxide(O