MilliporeSigma
  • Promethazine Hydrochloride Inhibits Ectopic Fat Cell Formation in Skeletal Muscle.
Promethazine Hydrochloride Inhibits Ectopic Fat Cell Formation in Skeletal Muscle.
The American journal of pathology (2017-09-19)
Takehiro Kasai, Masashi Nakatani, Naoki Ishiguro, Kinji Ohno, Naoki Yamamoto, Mitsuhiro Morita, Harumoto Yamada, Kunihiro Tsuchida, Akiyoshi Uezumi
ABSTRACT
Fatty degeneration of skeletal muscle leads to muscle weakness and loss of function. Preventing fatty degeneration in skeletal muscle is important, but no drug has been used clinically. In this study, we performed drug repositioning using human platelet-derived growth factor receptor α (PDGFRα)-positive mesenchymal progenitors that have been proved to be an origin of ectopic adipocytes in skeletal muscle. We found that promethazine hydrochloride (PH) inhibits adipogenesis in a dose-dependent manner without cell toxicity. PH inhibited expression of adipogenic markers and also suppressed phosphorylation of cAMP response-element binding protein, which was reported to be a primary regulator of adipogenesis. We established a mouse model of tendon rupture with intramuscular fat deposition and confirmed that emerged ectopic adipocytes are derived from PDGFRα
Sigma-Aldrich
Anti-Perilipin A/B antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution