MilliporeSigma
  • Size dependence of gold clusters with precise numbers of atoms in aerobic oxidation of d-glucose.
Size dependence of gold clusters with precise numbers of atoms in aerobic oxidation of d-glucose.
Nanoscale (2017-10-28)
Junying Zhang, Zhimin Li, Jiahui Huang, Chao Liu, Feng Hong, Kai Zheng, Gao Li
ABSTRACT
Size-dependence is an important factor in gold nanocatalysis. In this study, we explored the catalytic performance of atomically precise Au
Sigma-Aldrich
Phenylethyl mercaptan, ≥99%, FG