MilliporeSigma
  • MUC1 inhibition leads to decrease in PD-L1 levels via upregulation of miRNAs.
MUC1 inhibition leads to decrease in PD-L1 levels via upregulation of miRNAs.
Leukemia (2017-05-31)
A R Pyzer, D Stroopinsky, J Rosenblatt, E Anastasiadou, H Rajabi, A Washington, A Tagde, J-H Chu, M Coll, A L Jiao, L T Tsai, D E Tenen, L Cole, K Palmer, A Ephraim, R K Leaf, M Nahas, A Apel, M Bar-Natan, S Jain, M McMasters, L Mendez, J Arnason, B A Raby, F Slack, D Kufe, D Avigan
ABSTRACT
The PD-L1/PD-1 pathway is a critical component of the immunosuppressive tumor microenvironment in acute myeloid leukemia (AML), but little is known about its regulation. We investigated the role of the MUC1 oncoprotein in modulating PD-L1 expression in AML. Silencing of MUC1 in AML cell lines suppressed PD-L1 expression without a decrease in PD-L1 mRNA levels, suggesting a post-transcriptional mechanism of regulation. We identified the microRNAs miR-200c and miR-34a as key regulators of PD-L1 expression in AML. Silencing of MUC1 in AML cells led to a marked increase in miR-200c and miR-34a levels, without changes in precursor microRNA, suggesting that MUC1 might regulate microRNA-processing. MUC1 signaling decreased the expression of the microRNA-processing protein DICER, via the suppression of c-Jun activity. NanoString (Seattle, WA, USA) array of MUC1-silenced AML cells demonstrated an increase in the majority of probed microRNAs. In an immunocompetent murine AML model, targeting of MUC1 led to a significant increase in leukemia-specific T cells. In concert, targeting MUC1 signaling in human AML cells resulted in enhanced sensitivity to T-cell-mediated lysis. These findings suggest MUC1 is a critical regulator of PD-L1 expression via its effects on microRNA levels and represents a potential therapeutic target to enhance anti-tumor immunity.
Sigma-Aldrich
CRISPR/Cas9 Products and Services, Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.
Sigma-Aldrich
MISSION® esiRNA, targeting human MUC1