MilliporeSigma
  • Holistic Lipidomics of the Human Gut Phenotype Using Validated Ultra-High-Performance Liquid Chromatography Coupled to Hybrid Orbitrap Mass Spectrometry.
Holistic Lipidomics of the Human Gut Phenotype Using Validated Ultra-High-Performance Liquid Chromatography Coupled to Hybrid Orbitrap Mass Spectrometry.
Analytical chemistry (2017-10-21)
Lieven Van Meulebroek, Ellen De Paepe, Vicky Vercruysse, Beata Pomian, Simon Bos, Bruno Lapauw, Lynn Vanhaecke
ABSTRACT
As lipids are assigned a plethora of biological functions, it is evident that dysregulated lipid metabolism signifies a key element in many pathological conditions. With this rationale, this study presents a validated lipidomics platform to map the fecal lipidome, which integrates unique information about host-gut microbiome interactions, gastrointestinal functionality, and dietary patterns. This particular method accomplished coverage across all eight lipid categories: fatty acyls, glycerolipids, phosphoglycerolipids, polyketides, prenols, saccharolipids, sphingolipids, and sterols. Generic extraction of freeze-dried feces was achieved by solid-liquid extraction using methanol and methyl tert-butyl ether. Extracted components were separated by liquid chromatography, whereby the selected ethylene-bridged hybrid phenyl ultra-high-performance liquid chromatography stationary phase allowed fast separation of both individual lipid species and categories. Detection was achieved by high-resolution full-scan Q-Exactive Orbitrap mass spectrometry and covered a broad m/z scan range (67-2300 Da). Method validation was performed in a targeted fashion to evaluate the analytical performance across all lipid categories, revealing excellent linearity (R
Sigma-Aldrich
Dodecanedioic acid, 99%