MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Mouse models of nonalcoholic steatohepatitis in preclinical drug development.
Mouse models of nonalcoholic steatohepatitis in preclinical drug development.
Drug discovery today (2017-07-09)
Henrik H Hansen, Michael Feigh, Sanne S Veidal, Kristoffer T Rigbolt, Niels Vrang, Keld Fosgerau
ABSTRACT
Nonalcoholic fatty liver disease (NAFLD) has become the most common cause of chronic liver disease in the Western world. NAFLD is a complex spectrum of liver diseases ranging from benign hepatic steatosis to its more aggressive necroinflammatory manifestation, nonalcoholic steatohepatitis (NASH). NASH pathogenesis is multifactorial and risk factors are almost identical to those of the metabolic syndrome. This has prompted substantial efforts to identify novel drug therapies for correcting underlying metabolic deficits, and to prevent or alleviate hepatic fibrosis in NASH. Available mouse models of NASH address different aspects of the disease, have varying clinical translatability, and, therefore, also show different utility in drug discovery.
Sigma-Aldrich
Aliskiren