MilliporeSigma
  • Optimized light-inducible transcription in mammalian cells using Flavin Kelch-repeat F-box1/GIGANTEA and CRY2/CIB1.
Optimized light-inducible transcription in mammalian cells using Flavin Kelch-repeat F-box1/GIGANTEA and CRY2/CIB1.
Nucleic acids research (2017-10-19)
Jose R Quejada, Seon-Hye E Park, Daniel W Awari, Fan Shi, Hannah E Yamamoto, Fuun Kawano, Juergen C Jung, Masayuki Yazawa
ABSTRACT
Light-inducible systems allow spatiotemporal control of a variety of biological activities. Here, we report newly optimized optogenetic tools to induce transcription with light in mammalian cells, using the Arabidopsis photoreceptor Flavin Kelch-repeat F-box 1 (FKF1) and its binding partner GIGANTEA (GI) as well as CRY2/CIB1. By combining the mutagenesis of FKF1 with the optimization of a split FKF1/GI dimerized Gal4-VP16 transcriptional system, we identified constructs enabling significantly improved light-triggered transcriptional induction. In addition, we have improved the CRY2/CIB1-based light-inducible transcription with split construct optimization. The improvements regarding the FKF1/GI- and CRY2/CIB1-based systems will be widely applicable for the light-dependent control of transcription in mammalian cells.
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, clone TUB 2.1, purified from hybridoma cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Poly-DL-ornithine hydrobromide, mol wt 15,000-30,000
Scienceware® gel incubating tray, W × L 9.0 cm (3.5 in.) × 9.0 cm (3.5 in.)