MilliporeSigma
  • Hepatic protein tyrosine phosphatase receptor gamma links obesity-induced inflammation to insulin resistance.
Hepatic protein tyrosine phosphatase receptor gamma links obesity-induced inflammation to insulin resistance.
Nature communications (2017-11-29)
Xavier Brenachot, Giorgio Ramadori, Rafael M Ioris, Christelle Veyrat-Durebex, Jordi Altirriba, Ebru Aras, Sanda Ljubicic, Daisuke Kohno, Salvatore Fabbiano, Sophie Clement, Nicolas Goossens, Mirko Trajkovski, Sheila Harroch, Francesco Negro, Roberto Coppari
ABSTRACT
Obesity-induced inflammation engenders insulin resistance and type 2 diabetes mellitus (T2DM) but the inflammatory effectors linking obesity to insulin resistance are incompletely understood. Here, we show that hepatic expression of Protein Tyrosine Phosphatase Receptor Gamma (PTPR-γ) is stimulated by inflammation in obese/T2DM mice and positively correlates with indices of inflammation and insulin resistance in humans. NF-κB binds to the promoter of Ptprg and is required for inflammation-induced PTPR-γ expression. PTPR-γ loss-of-function lowers glycemia and insulinemia by enhancing insulin-stimulated suppression of endogenous glucose production. These phenotypes are rescued by re-expression of Ptprg only in liver of mice lacking Ptprg globally. Hepatic PTPR-γ overexpression that mimics levels found in obesity is sufficient to cause severe hepatic and systemic insulin resistance. We propose hepatic PTPR-γ as a link between obesity-induced inflammation and insulin resistance and as potential target for treatment of T2DM.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O127:B8, purified by phenol extraction