MilliporeSigma
  • Characterization of the estrogenic activities of zearalenone and zeranol in vivo and in vitro.
Characterization of the estrogenic activities of zearalenone and zeranol in vivo and in vitro.
The Journal of steroid biochemistry and molecular biology (2006-11-14)
Hitomi Takemura, Joong-Youn Shim, Kazutoshi Sayama, Airo Tsubura, Bao Ting Zhu, Kayoko Shimoi
ABSTRACT
In the present study, we compared the estrogenic activity of zearalenone (ZEN) and zeranol (ZOL) by determining their relative receptor binding affinities for human ERalpha and ERbeta and also by determining their uterotropic activity in ovariectomized female mice. ZOL displayed a much higher binding affinity for human ERalpha and ERbeta than ZEN did. The IC(50) values of ZEN and ZOL for binding to human ERalpha were 240.4 and 21.79nM, respectively, and the IC(50) values for binding to ERbeta were 165.7 and 42.76nM, respectively. In ovariectomized female ICR mice, s.c. administration of ZEN at doses >or=2mg/kg/day for 3 consecutive days significantly increased uterine wet weight compared with the control group, and administration of ZOL increased the uterine wet weight at lower doses (>or=0.5mg/kg/day for 3 days). Based on available X-ray crystal structures of human ERalpha and ERbeta, we have also conducted molecular modeling studies to probe the binding characteristics of ZEN and ZOL for human ERalpha and ERbeta. Our data revealed that ZEN and ZOL were able to occupy the active site of the human ERalpha and ERbeta in a strikingly similar manner as 17beta-estradiol, such that the phenolic rings of ZEN and ZOL occupied the same receptor region as occupied by the A-ring of 17beta-estradiol. The primary reason that ZOL and ZEN is less potent than 17beta-estradiol is likely because 17beta-estradiol could bind to the receptor pocket without significantly changing its conformation, while ZOL or ZEN would require considerable conformational alterations upon binding to the estrogen receptors (ERs).
Sigma-Aldrich
Zearalenone Ready Made Solution, 1 mg/mL in DMSO