MilliporeSigma
  • Comparison of Silodosin Monotherapy vs Silodosin With Tadalafil Add-on Therapy in Patients With Benign Prostatic Hyperplasia.
Comparison of Silodosin Monotherapy vs Silodosin With Tadalafil Add-on Therapy in Patients With Benign Prostatic Hyperplasia.
Urology (2017-04-23)
Takashi Yoshida, Hidefumi Kinoshita, Seiji Shimada, Makoto Taguchi, Tadashi Matsuda
ABSTRACT
To evaluate the efficacy and safety of add-on therapy with the phosphodiesterase type 5 inhibitor tadalafil for patients with lower urinary tract symptoms secondary to benign prostatic hyperplasia (LUTS/BPH) treated with the α We analyzed 103 patients with LUTS/BPH with an International Prostate Symptom Score (IPSS) of >8 after ≥4 weeks of silodosin treatment from April 2016 through December 2016 at Kori Hospital. The patients subsequently received silodosin 4.0 mg twice daily (monotherapy group) or silodosin 4.0 mg twice daily plus tadalafil 5.0 mg once daily (add-on therapy group) for 8 weeks. We assessed adverse events and evaluated the mean change from baseline to 8 weeks in the IPSS, Overactive Bladder Symptom Score (OABSS), maximum urine flow rate (Q Of 103 patients, 101 (98.1%) could continue medical treatment. The IPSS, OABSS, and Q Add-on therapy with tadalafil may be effective for patients with LUTS/BPH resistant to silodosin monotherapy.
Sigma-Aldrich
Silodosin, ≥98% (HPLC)