MilliporeSigma
  • High-throughput precision measurement of subcellular localization in single cells.
High-throughput precision measurement of subcellular localization in single cells.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2017-01-18)
Tyler J Burns, Andreas P Frei, Pier F Gherardini, Felice A Bava, Jake E Batchelder, Yuki Yoshiyasu, Julie M Yu, Amanda R Groziak, Samuel C Kimmey, Veronica D Gonzalez, Wendy J Fantl, Garry P Nolan
ABSTRACT
To quantify visual and spatial information in single cells with a throughput of thousands of cells per second, we developed Subcellular Localization Assay (SLA). This adaptation of Proximity Ligation Assay expands the capabilities of flow cytometry to include data relating to localization of proteins to and within organelles. We used SLA to detect the nuclear import of transcription factors across cell subsets in complex samples. We further measured intranuclear re-localization of target proteins across the cell cycle and upon DNA damage induction. SLA combines multiple single-cell methods to bring about a new dimension of inquiry and analysis in complex cell populations. © 2017 International Society for Advancement of Cytometry.
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection