MilliporeSigma
  • Supramolecular chiral self-assemblies of Gly-Pro dipeptides on metallic fcc(110) surfaces.
Supramolecular chiral self-assemblies of Gly-Pro dipeptides on metallic fcc(110) surfaces.
Faraday discussions (2017-08-03)
C Méthivier, H Cruguel, C-M Pradier, V Humblot
ABSTRACT
Adsorption of the Glycine-Proline (Gly-Pro) dipeptide has been investigated using surface science complementary techniques on Au(110) and Ag(110), showing some interesting differences both in the chemical form and surface organization of the adsorbed peptide. On Au(110), Gly-Pro mainly adsorbs in neutral form (COOH/NH
Sigma-Aldrich
Gly-Pro