MilliporeSigma
  • Chromatographic and Computational Assessment of Lipophilicity of New Anticancer Acetylenequinoline Derivatives.
Chromatographic and Computational Assessment of Lipophilicity of New Anticancer Acetylenequinoline Derivatives.
Journal of chromatographic science (2017-06-28)
Krzysztof Marciniec, Stanislaw Boryczka
ABSTRACT
The lipophilicity of a series of anticancer propargylquinoline derivatives is investigated using both chromatographic and computational methods. The parameters of the tested compounds' relative lipophilicity (logkw) are determined experimentally by the high-performance liquid chromatographic method (RP-HPLC, Accucore C18 column), using mixtures of acetonitrile and water as mobile phases. Mobile phase acetonitrile concentrations range between 50 and 80%. The logk values of the investigated compounds are linearly dependent upon the acetonitrile concentration. The analysis led to the calculation of the logkw parameter values for each of the tested compounds. The parameter logkw is discussed in terms of the relationship between structure and lipophilicity and consequently, transformed into the parameter logPHPLC using the calibration curve. The partition coefficients of the tested compounds (logPcalc) are also calculated by selected computer programs. A regression analysis and the sum of ranking differences are used to compare the lipophilic parameters of 15 acetylenequinoline derivatives, which were experimentally obtained (logPHPLC) and calculated using different mathematical methods (logPcalc). The 13C NMR spectra are used to examine the electronic relationships between properties and lipophilicity for the studied compounds. A regression study conducted on 15 compounds exhibits a linear correlation between lipophilicity and electronic properties, expressed as the 13C NMR chemical shift (R2 = 0.98).
Sigma-Aldrich
2-Chloroquinoline, 99%