MilliporeSigma
  • MCT1 and MCT4 Expression and Lactate Flux Activity Increase During White and Brown Adipogenesis and Impact Adipocyte Metabolism.
MCT1 and MCT4 Expression and Lactate Flux Activity Increase During White and Brown Adipogenesis and Impact Adipocyte Metabolism.
Scientific reports (2017-10-14)
Charlotte Petersen, Mette D Nielsen, Elise S Andersen, Astrid L Basse, Marie S Isidor, Lasse K Markussen, Birgitte M Viuff, Ian H Lambert, Jacob B Hansen, Stine F Pedersen
ABSTRACT
Adipose tissue takes up glucose and releases lactate, thereby contributing significantly to systemic glucose and lactate homeostasis. This implies the necessity of upregulation of net acid and lactate flux capacity during adipocyte differentiation and function. However, the regulation of lactate- and acid/base transporters in adipocytes is poorly understood. Here, we tested the hypothesis that adipocyte thermogenesis, browning and differentiation are associated with an upregulation of plasma membrane lactate and acid/base transport capacity that in turn is important for adipocyte metabolism. The mRNA and protein levels of the lactate-H
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)