MilliporeSigma
  • Short-term efficacy and safety of three different antiplatelet regimens in diabetic patients treated with primary percutaneous coronary intervention: a randomised study.
Short-term efficacy and safety of three different antiplatelet regimens in diabetic patients treated with primary percutaneous coronary intervention: a randomised study.
Kardiologia polska (2017-06-15)
Yang Liu, Hengliang Liu, Yibin Hao, Zhenxuan Hao, Guoying Geng, Wenjie Han, Qi Chen, Danli Wang, Lingzhi Liu, Kailong Jia, Yuxin Zhou
ABSTRACT
This study aimed to investigate the efficacy and safety of dual and triple antiplatelet therapy (DAPT and TAPT) in patients with diabetes and acute ST segment elevation myocardial infarction (D-STEMI), who had undergone primary percutaneous coronary intervention (PCI). We designed a phase IV, single-centre, randomised, double-blind, placebo-controlled study. The D-STEMI patients (n = 258) were randomly divided into three groups. Control group A (85 patients), was treated with aspirin and clopidogrel; group B (87 patients) received aspirin, clopidogrel, and tirofiban; and group C (86 patients) were treated with aspirin, ticagrelor, and tirofiban. Patients in all three groups received oral DAPT, and patients in groups B and C received intravenous tirofiban when primary PCI was performed. Compared to the findings in group A, the post-PCI Thrombolysis in Myocardial Infarction (TIMI) grade 3 blood flow in groups B and C increased significantly (TIMI grade 3 in groups A, B, C: 74%, 91%, and 98%, respectively; TIMI myocardial perfusion grade [TMPG] grade 3 in groups A, B, C: 59%, 86%, and 97%, respectively), and the incidence of major adverse cardiac events (MACE) decreased significantly (p < 0.05). Compared to the findings in group B, the rate of TMPG 3 in group C was significantly higher (p < 0.05) and the incidence of MACE was significantly lower (p < 0.05). Patients in group B exhibited minor bleeding; however, the incidence of mild to moderate bleeding in group C increased significantly (p < 0.05). TAPT effectively improved the TIMI blood flow and TMPG and reduced the occurrence of MACE. Ticagrelor was more effective than clopidogrel in TAPT; however, when using the combination of aspirin, ticagrelor, and tirofiban, close monitoring is required for possible bleeding complications.
Sigma-Aldrich
Tirofiban, ≥98.5% (HPLC)
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid