MilliporeSigma
  • Vinegar production from fruit concentrates: effect on volatile composition and antioxidant activity.
Vinegar production from fruit concentrates: effect on volatile composition and antioxidant activity.
Journal of food science and technology (2017-11-01)
Eduardo Coelho, Zlatina Genisheva, José Maria Oliveira, José António Teixeira, Lucília Domingues
ABSTRACT
Vinegar stands as a highly appreciated fermented food product due to several functional properties and multiple applications. This work focuses on vinegar production from fruit wines derived from fruit concentrates, to attain a food product with nutritional added value. Four fruit vinegars (orange, mango, cherry and banana), were produced and characterized, with total acidities of 5.3 ± 0.3% for orange, 5.6 ± 0.2% for mango, 4.9 ± 0.4% for cherry and 5.4 ± 0.4% for banana. Acetification showed impact on aroma volatiles, mainly related to oxidative reactions. Minor volatiles associated with varietal aroma were identified, monoterpenic alcohols in orange vinegar, esters in banana vinegar, C
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Geraniol, 98%
Sigma-Aldrich
3-Ethoxy-1-propanol, 97%