MilliporeSigma
  • Tuning and Predicting Mesh Size and Protein Release from Step Growth Hydrogels.
Tuning and Predicting Mesh Size and Protein Release from Step Growth Hydrogels.
Biomacromolecules (2017-08-30)
Matthew S Rehmann, Kelsi M Skeens, Prathamesh M Kharkar, Eden M Ford, Emanual Maverakis, Kelvin H Lee, April M Kloxin
ABSTRACT
Hydrogel-based depots are of growing interest for release of biopharmaceuticals; however, a priori selection of hydrogel compositions that will retain proteins of interest and provide desired release profiles remains elusive. Toward addressing this, in this work, we have established a new tool for the facile assessment of protein release from hydrogels and applied it to evaluate the effectiveness of mesh size estimations on predicting protein retention or release. Poly(ethylene glycol) (PEG)-based hydrogel depots were formed by photoinitiated step growth polymerization of four-arm PEG functionalized with norbornene (PEG-norbornene, 4% w/w to 20% w/w, M
Sigma-Aldrich
2,4,6-Trimethylbenzoyl chloride, 97%
Sigma-Aldrich
Dimethyl phenylphosphonite, 97%