MilliporeSigma
  • Electrolyte containing lithium cation in squaraine-sensitized solar cells: interactions and consequences for performance and charge transfer dynamics.
Electrolyte containing lithium cation in squaraine-sensitized solar cells: interactions and consequences for performance and charge transfer dynamics.
Physical chemistry chemical physics : PCCP (2017-10-07)
Vittoria Novelli, Nadia Barbero, Claudia Barolo, Guido Viscardi, Michel Sliwa, Frédéric Sauvage
ABSTRACT
By optimizing the lithium concentration in an electrolyte to 50 mmol L
Sigma-Aldrich
4-tert-Butylpyridine, 98%
Sigma-Aldrich
Valeronitrile, 99.5%
Sigma-Aldrich
Lithium iodide, AnhydroBeads, 99%