MilliporeSigma
  • Betanodavirus B2 protein triggers apoptosis and necroptosis in lung cancer cells that suppresses autophagy.
Betanodavirus B2 protein triggers apoptosis and necroptosis in lung cancer cells that suppresses autophagy.
Oncotarget (2017-12-08)
Hsuan-Wen Chiu, Yu-Chin Su, Jiann-Ruey Hong
ABSTRACT
The betanodavirus B2 protein targets the mitochondria and acts as a "death factor", but its effect on lung cancer cells is unknown. We examined the effect of the B2 protein on triggering apoptosis or necroptosis
Sigma-Aldrich
Necrostatin-1, ≥98% (HPLC)
Sigma-Aldrich
Pifithrin-α, ≥95% (HPLC), powder
Sigma-Aldrich
Polyethylenimine, branched, average Mw ~25,000 by LS, average Mn ~10,000 by GPC, branched