MilliporeSigma
  • Contribution of AcrAB-TolC to multidrug resistance in an Escherichia coli sequence type 131 isolate.
Contribution of AcrAB-TolC to multidrug resistance in an Escherichia coli sequence type 131 isolate.
International journal of antimicrobial agents (2017-07-12)
Sabine Schuster, Martina Vavra, Tobias M Schweigger, John W A Rossen, Yasufumi Matsumura, Winfried V Kern
ABSTRACT
Drug efflux by resistance-nodulation-cell division (RND)-type transporters, such as AcrAB-TolC of Escherichia coli, is an important resistance mechanism in Gram-negative bacteria; however, its contribution to multidrug resistance (MDR) in clinical isolates is poorly defined. We inactivated acrB of a sequence type 131 E. coli human isolate that showed high-level MDR, but had no mutations within the known efflux-associated local or global regulators. The resistance profile of the acrB deletion mutant revealed significantly increased susceptibility to drugs from seven antibiotic classes, including agents usually inactive against Gram-negative bacteria, notably the new oxazolidinone, tedizolid (512-fold enhanced susceptibility). AcrB deficiency reduced, but did not abolish, the efflux of dyes, which indicates the activity of at least one more efflux transporter. The findings demonstrate the efficacy of AcrAB-TolC-mediated broad-spectrum drug efflux, including agents primarily developed for Gram-positive pathogens, in a clinical isolate representative of a globally-emerging lineage. The results illustrate the need to develop molecules modified to impede their transport by AcrAB-TolC and its homologues and new efflux inhibitors.