MilliporeSigma
How a Transition-Metal(II) Chloride Interacts with a Eutectic AlCl
Chemistry, an Asian journal (2017-08-05)
Giridhar Pulletikurthi, Bernd Weidenfeller, Andriy Borodin, Jan C Namyslo, Frank Endres
ABSTRACT
Electrostatic interactions are characteristic of ionic liquids (ILs) and play a pivotal role in determining the formation of species when solutes are dissolved in them. The formation of new species/complexes has been investigated for certain ILs. However, such investigations have not yet focused on eutectic liquids, which are a promising class of ILs. These liquids (or liquid coordination complexes, LCCs) are rather new and are composed of cationic and anionic chloro complexes of metals. To date, these liquids have been employed as electrolytes to deposit metals and as solvents for catalysis. The present study deals with a liquid that is prepared by mixing a 1.2:1 mol ratio of AlCl
Sigma-Aldrich
1-Butylpyrrolidine, 98%