MilliporeSigma
  • Manganese(II) complexes with Bn-tpen as powerful catalysts of cyclohexene oxidation.
Manganese(II) complexes with Bn-tpen as powerful catalysts of cyclohexene oxidation.
Chemicke zvesti (2017-11-07)
Katarzyna Rydel-Ciszek, Maria Charczuk, Tomasz Pacześniak, Paweł Chmielarz
ABSTRACT
Manganese(II) complex [(Bn-tpen)Mn
Sigma-Aldrich
Cyclohexene oxide, 98%
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
2-Cyclohexen-1-one, ≥95%
Sigma-Aldrich
2-Cyclohexen-1-ol, 95%