MilliporeSigma
  • Globular domain structure and function of restriction-like-endonuclease LINEs: similarities to eukaryotic splicing factor Prp8.
Globular domain structure and function of restriction-like-endonuclease LINEs: similarities to eukaryotic splicing factor Prp8.
Mobile DNA (2017-11-21)
M Murshida Mahbub, Saiful M Chowdhury, Shawn M Christensen
ABSTRACT
R2 elements are a clade of early branching Long Interspersed Elements (LINEs). LINEs are retrotransposable elements whose replication can have profound effects on the genomes in which they reside. No crystal or EM structures exist for the reverse transcriptase (RT) and linker regions of LINEs. Using limited proteolysis as a probe for globular domain structure, we show that the protein encoded by the The protein encoded by RLE LINEs consists of two major globular domains. The larger of the two globular domain contains the RT, linker, and RLE and is similar to the large fragment of the spliceosomal protein Prp8. The similarities are suggestive of possible common ancestry.
Product Description
Supelco
SYPRO® Orange Protein Gel Stain