MilliporeSigma
  • Magnetized carbon nanotubes for visual detection of proteins directly in whole blood.
Magnetized carbon nanotubes for visual detection of proteins directly in whole blood.
Analytica chimica acta (2017-10-29)
Yan Huang, Yongqiang Wen, Kwaku Baryeh, Sunitha Takalkar, Michelle Lund, Xueji Zhang, Guodong Liu
ABSTRACT
The authors describe a magnetized carbon nanotube (MCNT)-based lateral flow strip biosensor for visual detection of proteins directly in whole blood avoiding complex purification and sample pre-treatments. MCNT were synthesized by coating Fe
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)
Sigma-Aldrich
Carbon nanotube, multi-walled, >90% carbon basis, D × L 110-170 nm × 5-9 μm
Sigma-Aldrich
Carbon nanotube, multi-walled, carboxylic acid functionalized, thin, extent of labeling: >8% carboxylic acid functionalized, avg. diam. × L 9.5 nm × 1.5 μm