MilliporeSigma
  • Highly sensitive method for the quantification of trans-linolenic acid isomers in trilinolenin of edible oils using an ionic liquid capillary column.
Highly sensitive method for the quantification of trans-linolenic acid isomers in trilinolenin of edible oils using an ionic liquid capillary column.
Journal of the science of food and agriculture (2017-04-04)
Qin Guo, Fan Jiang, Jing Jin, Qingpeng Li, Feng Wang, Qiang Wang, Yiming Ha
ABSTRACT
The polarities of linolenic acid isomers are very similar, and only a few studies to date have attempted to separate α-linolenic acid (ALA) isomers completely. The aim of this study was to fill this gap by developing and validating an accurate method for the analysis of ALA isomers in trilinolenin at 200, 220 and 240 °C using a gas chromatograph-flame ionization detector equipped with an SLB-IL111 capillary column. Results showed that eight ALA isomer standards were separated effectively using these optimized gas chromatographic conditions. The coefficient of determination was r The developed method has been successfully applied to the analysis of ALA isomers in heated pure trilinolenin as well as to trilinolenin in various edible oils, and the TALA isomerization pathways in heated trilinolenin were verified. © 2017 Society of Chemical Industry.
Sigma-Aldrich
Glyceryl trilinolenate, ≥97% (TLC), liquid