MilliporeSigma
  • High Quantum Yield Blue Emission from Lead-Free Inorganic Antimony Halide Perovskite Colloidal Quantum Dots.
High Quantum Yield Blue Emission from Lead-Free Inorganic Antimony Halide Perovskite Colloidal Quantum Dots.
ACS nano (2017-09-08)
Jian Zhang, Ying Yang, Hui Deng, Umar Farooq, Xiaokun Yang, Jahangeer Khan, Jiang Tang, Haisheng Song
ABSTRACT
Colloidal quantum dots (QDs) of lead halide perovskite have recently received great attention owing to their remarkable performances in optoelectronic applications. However, their wide applications are hindered from toxic lead element, which is not environment- and consumer-friendly. Herein, we utilized heterovalent substitution of divalent lead (Pb
Sigma-Aldrich
Cesium iodide, 99.9% trace metals basis
Sigma-Aldrich
Cesium chloride, 99.99% trace metals basis
Sigma-Aldrich
Cesium bromide, 99.9% trace metals basis
Sigma-Aldrich
Antimony(III) iodide, 98%
Sigma-Aldrich
Antimony(III) chloride, ReagentPlus®, 99%