MilliporeSigma
PADI2-Mediated Citrullination Promotes Prostate Cancer Progression.
Cancer research (2017-08-19)
Lin Wang, Guanhua Song, Xiang Zhang, Tingting Feng, Jihong Pan, Weiwen Chen, Muyi Yang, Xinnuo Bai, Yu Pang, Jindan Yu, Jinxiang Han, Bo Han
ABSTRACT
Onset of castration-resistance prostate cancer (CRPC) after long-term androgen deprivation therapy remains a major obstacle in the treatment of prostate cancer. The peptidylarginine deiminase PADI2 has been implicated in chronic inflammatory diseases and cancer. Here we show that
Sigma-Aldrich
MISSION® esiRNA, targeting human PADI2