MilliporeSigma
  • Molecular cloning, biochemical characterization, and expression analysis of two glutathione S-transferase paralogs from the big-belly seahorse (Hippocampus abdominalis).
Molecular cloning, biochemical characterization, and expression analysis of two glutathione S-transferase paralogs from the big-belly seahorse (Hippocampus abdominalis).
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology (2017-09-09)
M D Neranjan Tharuka, S D N K Bathige, Jehee Lee
ABSTRACT
Glutathione S-transferases (GSTs, EC 2.5.1.18) are important Phase II detoxifying enzymes that catalyze hydrophobic, electrophilic xenobiotic substance with the conjugation of reduced glutathione (GSH). In this study, GSTμ and GSTρ paralogs of GST in the big belly seahorse (Hippocampus abdominalis; HaGSTρ, HaGSTμ) were biochemically, molecularly, functionally characterized to determine their detoxification range and protective capacities upon different pathogenic stresses. HaGSTρ and HaGSTμ are composed of coding sequences of 681bp and 654bp, which encode proteins 225 and 217 amino acids, with predicted molecular masses of 26.06kDa and 25.74kDa respectively. Sequence analysis revealed that both HaGSTs comprise the characteristic GSH-binding site in the thioredoxin-like N-terminal domain and substrate binding site in the C-terminal domain. The recombinant HaGSTρ and HaGSTμ proteins catalyzed the model GST substrate 1-chloro-2, 4-dinitrobenzene (CDNB). Enzyme kinetic analysis revealed different K
Sigma-Aldrich
4-Nitrobenzyl chloride, 99%