MilliporeSigma
  • Dysregulated gene expressions of MEX3D, FOS and BCL2 in human induced-neuronal (iN) cells from NF1 patients: a pilot study.
Dysregulated gene expressions of MEX3D, FOS and BCL2 in human induced-neuronal (iN) cells from NF1 patients: a pilot study.
Scientific reports (2017-10-27)
Noriaki Sagata, Takahiro A Kato, Shin-Ichi Kano, Masahiro Ohgidani, Norihiro Shimokawa, Mina Sato-Kasai, Kohei Hayakawa, Nobuki Kuwano, Ashley M Wilson, Koko Ishizuka, Shiori Kato, Takeshi Nakahara, Makiko Nakahara-Kido, Daiki Setoyama, Yasunari Sakai, Shouichi Ohga, Masutaka Furue, Akira Sawa, Shigenobu Kanba
ABSTRACT
Direct conversion technique to produce induced-neuronal (iN) cells from human fibroblasts within 2 weeks is expected to discover unknown neuronal phenotypes of neuropsychiatric disorders. Here, we present unique gene expression profiles in iN cells from patients with neurofibromatosis type 1 (NF1), a single-gene multifaceted disorder with comparatively high co-occurrence of autism spectrum disorder (ASD). Microarray-based transcriptomic analysis on iN cells from male healthy controls and male NF1 patients (NF1-iN cells) revealed that 149 genes expressions were significantly different (110 upregulated and 39 downregulated). We validated that mRNA of MEX3D (mex-3 RNA binding family member D) was lower in NF1-iN cells by real-time PCR with 12 sex-mixed samples. In NF1-iN cells on day 14, higher expression of FOS mRNA was observed with lower expression of MEX3D mRNA. Interestingly, BCL2 mRNA was higher in NF1-iN cells on day 5 (early-period) but not on day 14. Our data suggest that aberrant molecular signals due to NF1 mutations may disturb gene expressions, a subset of which defines continuum of the neuronal phenotypes of NF1 with ASD. Further translational studies using induced pluripotent stem (iPS) cell-derived neuronal cells are needed to validate our preliminary findings especially confirming meanings of analysis using early-period iN cells.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mex3d
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Monoclonal Anti-β-Tubulin III antibody produced in mouse, clone SDL.3D10, ascites fluid