MilliporeSigma
  • Pharmacological analysis of the inhibition produced by moxonidine and agmatine on the vasodepressor sensory CGRPergic outflow in pithed rats.
Pharmacological analysis of the inhibition produced by moxonidine and agmatine on the vasodepressor sensory CGRPergic outflow in pithed rats.
European journal of pharmacology (2017-07-12)
Eloísa Rubio-Beltrán, Alejandro Labastida-Ramírez, Oswaldo Hernández-Abreu, Antoinette MaassenVanDenBrink, Carlos M Villalón
ABSTRACT
Calcitonin gene-related peptide (CGRP) plays a role in several (patho)physiological functions, and modulation of its release is considered a therapeutic target. In this respect, electrical spinal (T
Sigma-Aldrich
Hexamethonium chloride, solid (hygroscopic)
Sigma-Aldrich
BU224 hydrochloride, solid
Sigma-Aldrich
Moxonidine hydrochloride, ≥98%