MilliporeSigma
  • Activated spleen tyrosine kinase promotes malignant progression of oral squamous cell carcinoma via mTOR/S6 signaling pathway in an ERK1/2-independent manner.
Activated spleen tyrosine kinase promotes malignant progression of oral squamous cell carcinoma via mTOR/S6 signaling pathway in an ERK1/2-independent manner.
Oncotarget (2017-11-16)
Pan Gao, Xianghe Qiao, Haibin Sun, Yi Huang, Jie Lin, Longjiang Li, Xiaoyi Wang, Chunjie Li
ABSTRACT
Spleen tyrosine kinase (SYK), a non-receptor cytoplasmic tyrosine enzyme, is well known for its ability in certain pathways through immune receptors. Recently,
Sigma-Aldrich
Piceatannol, powder
Sigma-Aldrich
MISSION® esiRNA, targeting human SYK