MilliporeSigma
  • Construction of a hydrazone-linked chiral covalent organic framework-silica composite as the stationary phase for high performance liquid chromatography.
Construction of a hydrazone-linked chiral covalent organic framework-silica composite as the stationary phase for high performance liquid chromatography.
Journal of chromatography. A (2017-09-14)
Kai Zhang, Song-Liang Cai, Yi-Lun Yan, Zi-Hao He, Hui-Mei Lin, Xiao-Ling Huang, Sheng-Run Zheng, Jun Fan, Wei-Guang Zhang
ABSTRACT
Covalent organic frameworks (COFs), as an emerging class of crystalline porous organic polymers, have great potential for applications in chromatographic separation owning to their fascinating crystalline structures and outstanding properties. However, development of COF materials as novel stationary phases in high performance liquid chromatography (HPLC) is just in its infancy. Herein, we report the design and construction of a new hydrazone-linked chiral COF, termed BtaMth COF, from a chiral hydrazide building block (Mth) and present a one-pot synthetic method for the fabrication of BtaMth@SiO
Product Description
Supelco
Metconazole, PESTANAL®, analytical standard, mixture of stereoisomers