MilliporeSigma
  • Osm1 facilitates the transfer of electrons from Erv1 to fumarate in the redox-regulated import pathway in the mitochondrial intermembrane space.
Osm1 facilitates the transfer of electrons from Erv1 to fumarate in the redox-regulated import pathway in the mitochondrial intermembrane space.
Molecular biology of the cell (2017-08-18)
Sonya E Neal, Deepa V Dabir, Juwina Wijaya, Cennyana Boon, Carla M Koehler
ABSTRACT
Prokaryotes have aerobic and anaerobic electron acceptors for oxidative folding of periplasmic proteins. The mitochondrial intermembrane space has an analogous pathway with the oxidoreductase Mia40 and sulfhydryl oxidase Erv1, termed the mitochondrial intermembrane space assembly (MIA) pathway. The aerobic electron acceptors include oxygen and cytochrome
Sigma-Aldrich
3,3′-Dipropylthiadicarbocyanine iodide, suitable for fluorescence, ≥98.0% (TLC)