- Flexible PVDF membranes with exceptional robust superwetting surface for continuous separation of oil/water emulsions.
Flexible PVDF membranes with exceptional robust superwetting surface for continuous separation of oil/water emulsions.
Scientific reports (2017-10-28)
Zhu Xiong, Haibo Lin, Fu Liu, Peng Xiao, Ziyang Wu, Tiantian Li, Dehong Li
PMID29074888
ABSTRACT
Instability of superwetting surface is the stumbling block of flexible polymeric membranes for continuous separation of water-in-oil or oil-in-water emulsions. Manipulation of rigid superwetting nano-TiO