MilliporeSigma
  • Precise Composition Tailoring of Mixed-Cation Hybrid Perovskites for Efficient Solar Cells by Mixture Design Methods.
Precise Composition Tailoring of Mixed-Cation Hybrid Perovskites for Efficient Solar Cells by Mixture Design Methods.
ACS nano (2017-08-24)
Liang Li, Na Liu, Ziqi Xu, Qi Chen, Xindong Wang, Huanping Zhou
ABSTRACT
Mixed anion/cation perovskites absorber has been recently implemented to construct highly efficient single junction solar cells and tandem devices. However, considerable efforts are still required to map the composition-property relationship of the mixed perovskites absorber, which is essential to facilitate device design. Here we report the intensive exploration of mixed-cation perovskites in their compositional space with the assistance of a rational mixture design (MD) methods. Different from the previous linear search of the cation ratios, it is found that by employing the MD methods, the ternary composition can be tuned simultaneously following simplex lattice designs or simplex-centroid designs, which enable significantly reduced experiment/sampling size to unveil the composition-property relationship for mixed perovskite materials and to boost the resultant device efficiency. We illustrated the composition-property relationship of the mixed perovskites in multidimension and achieved an optimized power conversion efficiency of 20.99% in the corresponding device. Moreover, the method is demonstrated to be feasible to help adjust the bandgap through rational materials design, which can be further extended to other materials systems, not limited in polycrystalline perovskites films for photovoltaic applications only.
Sigma-Aldrich
Chlorobenzene, suitable for HPLC, 99.9%
Sigma-Aldrich
4-tert-Butylpyridine, 98%