MilliporeSigma
  • A novel CCR-2/TLR-2 triggered signaling in murine peritoneal macrophages intensifies bacterial (Staphylococcus aureus) killing by reactive oxygen species through TNF-R1.
A novel CCR-2/TLR-2 triggered signaling in murine peritoneal macrophages intensifies bacterial (Staphylococcus aureus) killing by reactive oxygen species through TNF-R1.
Immunology letters (2017-07-25)
Ajeya Nandi, Biswadev Bishayi
ABSTRACT
Macrophages are remarkably versatile in their ability to recognize and respond to a wide range of stimuli by expressing a variety of surface and intracellular receptors and triggering multiple signal transduction pathways. The onset of microbial infection is primarily determined by the initial contacts made by the microbes with the host macrophages. Although there prevail a relationship between the chemokine receptor and Toll like receptors during disease, particularly TLR-2 and CCR-2 signaling interdependence on each other has not been yet investigated during acute staphylococcal infection. Thus, the present study was aimed to trace possible interaction between CCR-2 and TLR-2 in peritoneal macrophages during acute Staphylococcus aureus infection. We found that neutralization of CCR-2 attenuates TLR-2 expression and restricts S. aureus burden but TLR-2 neutralization augments CCR-2 expression in macrophages, along with compromised host-derived reactive oxygen species production. S. aureus infection to CCR-2 intact but TLR-2 neutralized macrophages triggered production of IL-1β, TNF-α, IL-6, IFN-γ, MCP-1 and expression of iNOS, TNFR-1 and GPx with concomitant decrease in IL-10 production. Further, study with NG-monomethyl-l-arginine (L-NMMA) [iNOS blocker] and buthionine sulfoximine (BSO) [GPx blocker] revealed that S. aureus infection enhanced TLR-2 expression in CCR-2 intact and TLR-2 neutralized macrophages possibly via iNOS and TNFR-1 up regulation and GPx down regulation. Overall, our data indicate that targeting CCR-2 with neutralizing antibody in the early phase of S. aureus infection could restrict excessive inflammation with less compromised bacterial killing. It certainly would be a therapeutic strategy in S. aureus induced inflammatory and infective diseases.
Sigma-Aldrich
L-Buthionine-sulfoximine, ≥97% (TLC)
Sigma-Aldrich
NG-Methyl-L-arginine acetate salt, ≥98% (TLC)