MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • SG2NA is a regulator of endoplasmic reticulum (ER) homeostasis as its depletion leads to ER stress.
SG2NA is a regulator of endoplasmic reticulum (ER) homeostasis as its depletion leads to ER stress.
Cell stress & chaperones (2017-06-22)
Buddhi Prakash Jain, Shweta Pandey, Nikhat Saleem, Goutam K Tanti, Shalini Mishra, Shyamal K Goswami
ABSTRACT
SG2NA belongs to a three-member striatin subfamily of WD40 repeat superfamily of proteins. It has multiple protein-protein interaction domains involved in assembling supramolecular signaling complexes. Earlier, we had demonstrated that there are at least five variants of SG2NA generated by alternative splicing, intron retention, and RNA editing. Such versatile and dynamic mode of regulation implicates it in tissue development. In order to shed light on its role in cell physiology, total proteome analysis was performed in NIH3T3 cells depleted of 78 kDa SG2NA, the only isoform expressing therein. A number of ER stress markers were among those modulated after knockdown of SG2NA. In cells treated with the ER stressors thapsigargin and tunicamycin, expression of SG2NA was increased at both mRNA and protein levels. The increased level of SG2NA was primarily in the mitochondria and the microsomes. A mouse injected with thapsigargin also had an increase in SG2NA in the liver but not in the brain. Cell cycle analysis suggested that while loss of SG2NA reduces the level of cyclin D1 and retains a population of cells in the G1 phase, concurrent ER stress facilitates their exit from G1 and traverse through subsequent phases with concomitant cell death. Thus, SG2NA is a component of intrinsic regulatory pathways that maintains ER homeostasis.
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
MISSION® esiRNA, targeting human STRN3
Sigma-Aldrich
OptiPrep Density Gradient Medium, used for cell and subcellular organelle isolation