MilliporeSigma
  • Variation in developmental trajectories of physiological and somatic traits in a common songbird approaching fledging.
Variation in developmental trajectories of physiological and somatic traits in a common songbird approaching fledging.
The Journal of experimental biology (2017-09-17)
Allison Cornell, Tony D Williams
ABSTRACT
In avian species, little is known about the development of physiological traits in the days preceding fledging, a critical life history transition marked by a high mortality rate. Developmental trajectory during this period may be flexible based on ecological context or hardwired, with potential costs for variation in growth in the form of oxidative stress. Patterns in development are likely to relate to variation in life history, for which seabirds and aerial insectivores have been well studied, while our focal species is a grassland ground forager, the European starling (
Sigma-Aldrich
Drabkin′s reagent, for the determination of hemoglobin